Merck
  • Impact of anticoagulation on ionic and nonionic contrast media effect on thrombogenesis and fibrinolysis: The PEPCIT study.
Impact of anticoagulation on ionic and nonionic contrast media effect on thrombogenesis and fibrinolysis: The PEPCIT study.
Catheterization and cardiovascular interventions : official journal of the Society for Cardiac Angiography & Interventions (2011-05-28)
Anne Bellemain-Appaix, Farzin Beygui, Claude Lesty, Satya Gupta, Johanne Silvain, Claude Le Feuvre, Guillaume Cayla, Yann Allali, Gilles Montalescot, Jean-Philippe Collet
ABSTRACT
The effect of ionic low osmolar contrast media (ICM) and nonionic iso-osmolar CM (NICM) on acute thrombotic complications of percutaneous coronary intervention (PCI) is subject to controversies possibly related to a potential interaction with anticoagulation regimens. We sought to compare physical and morphological properties of fibrin clots made in the presence of ioxaglate (ICM), iodixanol (NICM) versus control and to evaluate the effect of four anticoagulants used in PCI. Maximum platelet aggregation (MPA%), maximum elastic modulus (EM, dyne/cm(2) ) fiber density (n/10(-5) /μm(2) ), and lysis front velocity (nm/sec) of fibrin rich clot (FRC) were measured simultaneously using peripheral blood from 12 patients undergoing elective PCI. We compared the effects of adding iodixanol or ioxaglate or saline (control) to blood with enoxaparin, unfractionated heparin, fondaparinux, and bivalirudin. Iodixanol and ioxaglate led to nonsignificant reduction in MPA compared to control (33.6% ± 16.9%, 28.2% ± 18.9%, and 40.7% ± 13.9%, respectively, P = ns). Fibrin formed with iodixanol was stiffer (42.7 ± 41.9, 18.7 ± 3.7, and 15.9 ± 9 dyne/cm(2) , P < 0.01) and displayed more fibrin fibers (1089 ± 175, 260 ± 108, and 456 ± 131 n/10(-5) /μm(2) , respectively, P < 0.01) than with ioxaglate or control. This resulted in a profound reduction in the lysis front velocity (191 ± 95, 261 ± 112, and 360 ± 153 nm/sec). None of the four anticoagulants displayed any significant interaction on the effect of contrast media. The prothrombogenic effect of iodixanol is related primarily to an increase in fibrin stiffness with subsequent delayed fibrinolysis, something not seen with ioxaglate. Anticoagulation does not appear to have any impact on this fibrin clot abnormalities.
Product Description
Ioxaglic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ioxaglic acid, United States Pharmacopeia (USP) Reference Standard