Merck
  • Excellence in exotics: Case report: urolithiasis in a female Guinea pig.
Excellence in exotics: Case report: urolithiasis in a female Guinea pig.
Compendium (Yardley, PA) (2013-03-28)
Heidi L Hoefer
ABSTRACT
A 2-year-old, intact female guinea pig presented to the clinic for a history of squealing and straining to urinate for 2 days. The husbandry of this animal was acceptable, and the diet consisted of commercial alfalfa-based guinea pig pellets, alfalfa hay, a variety of vegetables, and a water-soluble vitamin C supplement.
Product Description
Meloxicam, European Pharmacopoeia (EP) Reference Standard