Merck
  • Identification of priority pharmaceuticals in the water environment of China.
Identification of priority pharmaceuticals in the water environment of China.
Chemosphere (2012-05-18)
Qian Sui, Bin Wang, Wentao Zhao, Jun Huang, Gang Yu, Shubo Deng, Zhaofu Qiu, Shuguang Lu
ABSTRACT
In recent years, increasing attention has been paid to the trace-level contamination of pharmaceuticals in the water environment all over the world. Considering a large number of pharmaceuticals used, it is crucial to establish a priority list of pharmaceuticals that should be monitored and/or treated first. In the present study, we developed a ranking system based on the pharmaceutical consumption, removal performance in the wastewater treatment plants (WWTPs) and potential ecological effects, and applied to the situation of China. 39 pharmaceuticals, which had available consumption data and also been reported previously in the WWTPs of China, were selected as candidate pharmaceuticals. Among them, seventeen pharmaceuticals were considered as priority pharmaceuticals, out of which, erythromycin, diclofenac acid and ibuprofen, had the high priority. Compared with other literatures, we found that some pharmaceuticals given concerns to globally should also be included in the priority list in China; while some pharmaceuticals, not mentioned in other literatures, such as cefalexin, ketoconazole, should be also given prior consideration in China. Among all the therapeutic classes, antibiotics, which were grossly abused in China, contributed the most to the priority pharmaceuticals. However, priority antibiotics accounted for only 32% of candidate antibiotics, while 71% and 100% of the candidate anti-inflammatory and antilipidemic respectively were identified as the priority pharmaceuticals, indicating that antibiotics might be overanxiously considered in the previous studies on their behaviors in the WWTPs of China.
Product Description
Supelco
Cefalexin, VETRANAL®, analytical standard
Sigma-Aldrich
Cephalexin hydrate, first-generation cephalosporin antibiotic
Cephalexin monohydrate, European Pharmacopoeia (EP) Reference Standard