Merck
  • Polyelectrolyte-coated alginate microspheres as drug delivery carriers for dexamethasone release.
Polyelectrolyte-coated alginate microspheres as drug delivery carriers for dexamethasone release.
Drug delivery (2009-07-18)
R D Jayant, M J McShane, R Srivastava
ABSTRACT
Alginate microspheres loaded with dexamethasone were prepared by the droplet generator technique. Important parameters affecting drug release, including initial drug content, the type of polyelectrolyte coating, and a combination of different ratios of coated and uncoated microspheres were investigated to achieve in vitro dexamethasone delivery with approximately zero order release kinetics, releasing up to 100% of entrapped drug within 1 month, wherein dexamethasone released at a steady rate of 4.83 microg/day after an initial burst release period. These findings imply that these polyelectrolyte-coated alginate microspheres show promise as release systems to improve biocompatibility and prolong lifetime of implantable glucose sensors.
Sigma-Aldrich
Poly[(isobutylene-alt-maleic acid, ammonium salt)-co-(isobutylene-alt-maleic anhydride)], average Mw ~60,000
Sigma-Aldrich
Poly(isobutylene-alt-maleic anhydride), average Mw ~6,000, 12-200 mesh (85%)