Merck
  • Protease activation, pancreatic leakage, and inflammation in acute pancreatitis: differences between mild and severe cases and changes over the first three days.
Protease activation, pancreatic leakage, and inflammation in acute pancreatitis: differences between mild and severe cases and changes over the first three days.
Pancreatology : official journal of the International Association of Pancreatology (IAP) ... [et al.] (2008-10-14)
S Regnér, J Manjer, S Appelros, C Hjalmarsson, J Sadic, A Borgström
ABSTRACT
The pathophysiology of acute pancreatitis (AP) may be studied using markers of protease activation (active carboxypeptidase B (aCAP), the activation peptide of carboxypeptidase B (CAPAP)), leakage of pancreatic enzymes (trypsinogen-2, procarboxypeptidase B (proCAP), amylase), and inflammation (monocyte chemoattractant protein-1 (MCP-1), CRP). This prospective study included 140 cases of AP. Mild (n = 124) and severe (n = 16) cases were compared with respect to serum levels of trypsinogen-2, proCAP, amylase, aCAP, CAPAP (serum/urine), MCP-1 (serum/urine) and CRP on days 1, 2 and 3 from onset of symptoms. All patients with information on all 3 days were included in a time-course analysis (n = 44-55, except amylase: n = 27). High levels in severe versus mild cases were seen for trypsinogen-2, CAPAP in serum and urine, and MCP-1 in serum on days 1-3. No differences were seen for proCAP, amylase and aCAP. MCP-1 in urine was significantly elevated on day 1-2, and CRP on day 2-3. CAPAP and MCP-1 levels peaked early and stayed elevated for 48 h in serum. Protease activation and inflammation are early events in AP, with high levels of these markers within 24 h. Protease activation declines after 48 h, whereas inflammation is present for a longer time.
Sigma-Aldrich
Carboxypeptidase B from human pancreas, 50-55 units/mg protein carboxypeptidase B
Sigma-Aldrich
Carboxypeptidase B from porcine pancreas, lyophilized powder