Merck
  • SARS-CoV-2 spike S1 subunit protein-mediated increase of beta-secretase 1 (BACE1) impairs human brain vessel cells.
SARS-CoV-2 spike S1 subunit protein-mediated increase of beta-secretase 1 (BACE1) impairs human brain vessel cells.
Biochemical and biophysical research communications (2022-08-16)
Ji-Young Choi, Jung Hyun Park, Chulman Jo, Kyung-Chang Kim, Young Ho Koh
ABSTRACT
Increasing evidence suggests incomplete recovery of COVID-19 patients, who continue to suffer from cardiovascular diseases, including cerebral vascular disorders (CVD) and neurological symptoms. Recent findings indicate that some of the damaging effects of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) infection, especially in the brain, may be induced by the spike protein, leading to the disruption of the initial blood-brain barrier (BBB). SARS-CoV-2-infected cells and animals exhibit age-dependent pathogenesis. In this study, we identified endothelial BACE1 as a critical mediator of BBB disruption and cellular senescence induced by the SARS-CoV-2 spike S1 subunit protein. Increased BACE1 in human brain microvascular endothelial cells (HBMVEC) decreases the levels of tight junction proteins, including ZO-1, occludin, and claudins. Moreover, BACE1 overexpression leads to the accumulation of p16 and p21, typical hallmarks of cellular senescence. Our findings show that the SARS-CoV-2 spike S1 subunit protein upregulated BACE1 expression in HBMVECs, causing endothelial leakage. In addition, the SARS-CoV-2 spike S1 subunit protein induced p16 and p21 expression, indicating BACE1-mediated cellular senescence, confirmed by β-Gal staining in HBMVECs. In conclusion, this study demonstrated that BACE1-mediated endothelial cell damage and senescence may be linked to CVD after COVID-19 infection.
Sigma-Aldrich
In Vitro Vascular Permeability Assay (24-well), This In Vitro Vascular Permeability Assay kit employs a 24-well plate and provides an efficient system for evaluating the effects of chemicals & drug compounds on endothelial cell adsorption, transport & permeability.
Sigma-Aldrich
β-Secretase Inhibitor IV, β-Secretase Inhibitor IV, CAS 797035-11-1, is a cell-permeable inhibitor that binds to BACE-1 active site and blocks its proteolytic activity (IC₅₀ = 15 nM for human BACE-1).
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid