Merck
  • δ-Catenin Participates in EGF/AKT/p21Waf Signaling and Induces Prostate Cancer Cell Proliferation and Invasion.
δ-Catenin Participates in EGF/AKT/p21Waf Signaling and Induces Prostate Cancer Cell Proliferation and Invasion.
International journal of molecular sciences (2021-06-03)
Yingjie Shen, Hyoung Jae Lee, Rui Zhou, Hangun Kim, Gen Chen, Young-Chang Cho, Kwonseop Kim
ABSTRACT
Prostate cancer (PCa) is the second most leading cause of death in males. Our previous studies have demonstrated that δ-catenin plays an important role in prostate cancer progression. However, the molecular mechanism underlying the regulation of δ-catenin has not been fully explored yet. In the present study, we found that δ-catenin could induce phosphorylation of p21Waf and stabilize p21 in the cytoplasm, thus blocking its nuclear accumulation for the first time. We also found that δ-catenin could regulate the interaction between AKT and p21, leading to phosphorylation of p21 at Thr-145 residue. Finally, EGF was found to be a key factor upstream of AKT/δ-catenin/p21 for promoting proliferation and metastasis in prostate cancer. Our findings provide new insights into molecular controls of EGF and the development of potential therapeutics targeting δ-catenin to control prostate cancer progression.
Sigma-Aldrich
Leupeptin, BioUltra, microbial, ≥95% (HPLC)
Sigma-Aldrich
Aprotinin from bovine lung, saline solution, 3-7 TIU/mg protein
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid