Merck
  • Maternal N-Carbamylglutamate Supplementation during Early Pregnancy Enhances Embryonic Survival and Development through Modulation of the Endometrial Proteome in Gilts.
Maternal N-Carbamylglutamate Supplementation during Early Pregnancy Enhances Embryonic Survival and Development through Modulation of the Endometrial Proteome in Gilts.
The Journal of nutrition (2015-08-21)
Jinlong Zhu, Xiangfang Zeng, Qian Peng, Shenming Zeng, Haiyi Zhao, Hexiao Shen, Shiyan Qiao
ABSTRACT
Early pregnancy loss is a major concern in humans and animals. N-carbamylglutamate (NCG) has been found to enhance embryonic survival during early pregnancy in rats. However, little is known about the key factors in the endometrium involved in the improvement of embryonic implantation and development induced by maternal NCG supplementation. Our objectives were to investigate whether NCG supplementation during early gestation enhanced embryonic survival and development in gilts and to uncover the related factors using the approach of endometrium proteome analysis with isobaric tags for relative and absolute quantification (iTRAQ). Uteruses and embryos/fetuses were obtained on days 14 and 28 of gestation from gilts fed a basal diet that was or was not supplemented with 0.05% NCG. The iTRAQ-based quantitative proteomics approach was performed to explore the endometrium proteome altered by NCG supplementation. Maternal NCG supplementation significantly increased the number of total fetuses and live fetuses on day 28 of gestation by 1.32 and 1.29, respectively (P < 0.05), with a significant decrease in embryonic mortality (P < 0.05). iTRAQ results indicated that a total of 59 proteins showed at least 2-fold differences (P < 0.05), including 52 proteins that were present at higher abundance and 7 proteins present at lower abundance in NCG-supplemented gilts. The differentially expressed proteins primarily are involved in cell adhesion, energy metabolism, lipid metabolism, protein metabolism, antioxidative stress, and immune response. On day 14 of gestation, several proteins closely related to embryonic implantation and development, such as integrin-αv, integrin-β3, talin, and endothelial nitric oxide synthase, were upregulated (3.7-, 4.1-, 2.4-, and 5.4-fold increases, respectively) by NCG supplementation. To our knowledge, our results provide the first evidence that altered abundance of the endometrial proteome induced by NCG supplementation is highly associated with the improvement of embryonic survival and development in gilts.
Sigma-Aldrich
Nitrite/Nitrate Assay Kit, colorimetric, used for detection of nitric oxide metabolites