Merck
  • Small-molecule scaffolds for CYP51 inhibitors identified by high-throughput screening and defined by X-ray crystallography.
Small-molecule scaffolds for CYP51 inhibitors identified by high-throughput screening and defined by X-ray crystallography.
Antimicrobial agents and chemotherapy (2007-09-12)
Larissa M Podust, Jens P von Kries, Ali Nasser Eddine, Youngchang Kim, Liudmila V Yermalitskaya, Ronald Kuehne, Hugues Ouellet, Thulasi Warrier, Markus Alteköster, Jong-Seok Lee, Jörg Rademann, Hartmut Oschkinat, Stefan H E Kaufmann, Michael R Waterman
ABSTRACT
Sterol 14alpha-demethylase (CYP51), a major checkpoint in membrane sterol biosynthesis, is a key target for fungal antibiotic therapy. We sought small organic molecules for lead candidate CYP51 inhibitors. The changes in CYP51 spectral properties following ligand binding make CYP51 a convenient target for high-throughput screening technologies. These changes are characteristic of either substrate binding (type I) or inhibitor binding (type II) in the active site. We screened a library of 20,000 organic molecules against Mycobacterium tuberculosis CYP51 (CYP51(Mt)), examined the top type I and type II binding hits for their inhibitory effects on M. tuberculosis in broth culture, and analyzed them spectrally for their ability to discriminate between CYP51(Mt) and two reference M. tuberculosis CYP proteins, CYP130 and CYP125. We determined the binding mode for one of the top type II hits, alpha-ethyl-N-4-pyridinyl-benzeneacetamide (EPBA), by solving the X-ray structure of the CYP51(Mt)-EPBA complex to a resolution of 1.53 A. EPBA binds coordinately to the heme iron in the CYP51(Mt) active site through a lone pair of nitrogen electrons and also through hydrogen bonds with residues H259 and Y76, which are invariable in the CYP51 family, and hydrophobic interactions in a phylum- and/or substrate-specific cavity of CYP51. We also identified a second compound with structural and binding properties similar to those of EPBA, 2-(benzo[d]-2,1,3-thiadiazole-4-sulfonyl)-2-amino-2-phenyl-N-(pyridinyl-4)-acetamide (BSPPA). The congruence between the geometries of EPBA and BSPPA and the CYP51 binding site singles out EPBA and BSPPA as lead candidate CYP51 inhibitors with optimization potential for efficient discrimination between host and pathogen enzymes.
Sigma-Aldrich
Estriol, meets USP testing specifications
Sigma-Aldrich
4,4′-Dihydroxybenzophenone, 99%
Sigma-Aldrich
4-Phenylimidazole, 97%
Sigma-Aldrich
Estriol, ≥97%