Merck
  • Monocarboxylate transporter 1 in Schwann cells contributes to maintenance of sensory nerve myelination during aging.
Monocarboxylate transporter 1 in Schwann cells contributes to maintenance of sensory nerve myelination during aging.
Glia (2019-08-28)
Mithilesh Kumar Jha, Youngjin Lee, Katelyn A Russell, Fang Yang, Raha M Dastgheyb, Pragney Deme, Xanthe H Ament, Weiran Chen, Ying Liu, Yun Guan, Michael J Polydefkis, Ahmet Hoke, Norman J Haughey, Jeffrey D Rothstein, Brett M Morrison
ABSTRACT
Schwann cell (SC)-specific monocarboxylate transporter 1 (MCT1) knockout mice were generated by mating MCT1 f/f mice with myelin protein zero (P0)-Cre mice. P0-Cre+/- , MCT1 f/f mice have no detectable early developmental defects, but develop hypomyelination and reduced conduction velocity in sensory, but not motor, peripheral nerves during maturation and aging. Furthermore, reduced mechanical sensitivity is evident in aged P0-Cre+/- , MCT1 f/f mice. MCT1 deletion in SCs impairs both their glycolytic and mitochondrial functions, leading to altered lipid metabolism of triacylglycerides, diacylglycerides, and sphingomyelin, decreased expression of myelin-associated glycoprotein, and increased expression of c-Jun and p75-neurotrophin receptor, suggesting a regression of SCs to a less mature developmental state. Taken together, our results define the contribution of SC MCT1 to both SC metabolism and peripheral nerve maturation and aging.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Avanti
C12 Lactosyl(β) Ceramide (d18:1/12:0), Avanti Research - A Croda Brand 860545P, powder
Avanti
16:0 cholesteryl-d7 ester, Avanti Research - A Croda Brand 700149P, powder