Merck
  • Engineering peptide-targeted liposomal nanoparticles optimized for improved selectivity for HER2-positive breast cancer cells to achieve enhanced in vivo efficacy.
Engineering peptide-targeted liposomal nanoparticles optimized for improved selectivity for HER2-positive breast cancer cells to achieve enhanced in vivo efficacy.
Journal of controlled release : official journal of the Controlled Release Society (2020-04-11)
Baksun Kim, Jaeho Shin, Junmin Wu, David T Omstead, Tanyel Kiziltepe, Laurie E Littlepage, Basar Bilgicer
ABSTRACT
Here, we report rationally engineered peptide-targeted liposomal doxorubicin nanoparticles that have an enhanced selectivity for HER2-positive breast tumor cells with high purity, reproducibility, and precision in controlling stoichiometry of targeting peptides. To increase HER2-positive tumor cell selective drug delivery, we optimized the two most important design parameters, peptide density and linker length, via systematic evaluations of their effects on both in vitro cellular uptake and in vivo tumor accumulation and cellular uptake. The optimally designed nanoparticles were finally evaluated for their tumor inhibition efficacy using in vivo MMTV-neu transplantation mouse model. In vitro, we demonstrated that ~1% peptide density and EG8 linker were optimal parameters for targeted nanoparticle formulations to enhance HER2-positive cancer cellular uptake while preventing non-selectivity. In vivo results demonstrated that at 0.5% peptide density, enhancement of tumor cell uptake over non-targeted nanoparticles was ~2.7 fold and ~3.4 fold higher for targeted nanoparticles with EG8 and EG18 linker, respectively, while their accumulation levels at tumor tissue were similar to the non-targeted nanoparticles. These results were consistent with in vivo efficacy outcomes that ~90% tumor growth inhibition was achieved by Dox-loaded HER2 receptor targeted nanoparticles, TNPHER2pep, over control while all nanoparticle formulations minimized overall systemic toxicity relative to free Dox. This study highlights the significance of understanding and optimizing the effects of liposomal nanoparticle design parameters for enhancement of tumor selectivity to achieve improved in vivo therapeutic outcomes.
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
Triisopropylsilane, 98%
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand 880120C
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand
Avanti
16:0 Glutaryl PE, Avanti Research - A Croda Brand 870245P, powder
Sigma-Aldrich
1,2-Distearoyl-sn-glycero-3-phosphoethanolamine, ≥99%
Avanti
18:0 PC (DSPC), Avanti Research - A Croda Brand
Avanti
18:0 PC (DSPC), Avanti Research - A Croda Brand