Merck
  • Trisomy of human chromosome 21 enhances amyloid-β deposition independently of an extra copy of APP.
Trisomy of human chromosome 21 enhances amyloid-β deposition independently of an extra copy of APP.
Brain : a journal of neurology (2018-06-27)
Frances K Wiseman, Laura J Pulford, Chris Barkus, Fan Liao, Erik Portelius, Robin Webb, Lucia Chávez-Gutiérrez, Karen Cleverley, Sue Noy, Olivia Sheppard, Toby Collins, Caroline Powell, Claire J Sarell, Matthew Rickman, Xun Choong, Justin L Tosh, Carlos Siganporia, Heather T Whittaker, Floy Stewart, Maria Szaruga, Michael P Murphy, Kaj Blennow, Bart de Strooper, Henrik Zetterberg, David Bannerman, David M Holtzman, Victor L J Tybulewicz, Elizabeth M C Fisher
ABSTRACT
Down syndrome, caused by trisomy of chromosome 21, is the single most common risk factor for early-onset Alzheimer's disease. Worldwide approximately 6 million people have Down syndrome, and all these individuals will develop the hallmark amyloid plaques and neurofibrillary tangles of Alzheimer's disease by the age of 40 and the vast majority will go on to develop dementia. Triplication of APP, a gene on chromosome 21, is sufficient to cause early-onset Alzheimer's disease in the absence of Down syndrome. However, whether triplication of other chromosome 21 genes influences disease pathogenesis in the context of Down syndrome is unclear. Here we show, in a mouse model, that triplication of chromosome 21 genes other than APP increases amyloid-β aggregation, deposition of amyloid-β plaques and worsens associated cognitive deficits. This indicates that triplication of chromosome 21 genes other than APP is likely to have an important role to play in Alzheimer's disease pathogenesis in individuals who have Down syndrome. We go on to show that the effect of trisomy of chromosome 21 on amyloid-β aggregation correlates with an unexpected shift in soluble amyloid-β 40/42 ratio. This alteration in amyloid-β isoform ratio occurs independently of a change in the carboxypeptidase activity of the γ-secretase complex, which cleaves the peptide from APP, or the rate of extracellular clearance of amyloid-β. These new mechanistic insights into the role of triplication of genes on chromosome 21, other than APP, in the development of Alzheimer's disease in individuals who have Down syndrome may have implications for the treatment of this common cause of neurodegeneration.
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Alzheimer Precursor Protein A4 Antibody, clone 22C11, Biotin Conj. | MAB348B, clone 22C11, from mouse, biotin conjugate
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution