Merck
  • Structural characterization of resistant starch isolated from Laird lentils (Lens culinaris) seeds subjected to different processing treatments.
Structural characterization of resistant starch isolated from Laird lentils (Lens culinaris) seeds subjected to different processing treatments.
Food chemistry (2018-05-23)
Zhen Ma, Xiuxiu Yin, Xinzhong Hu, Xiaoping Li, Liu Liu, Joyce I Boye
ABSTRACT
This work focused on the structural characterization of resistant starch from untreated (UL-RS), germinated (GL-RS), fermented (FL-RS), microwaved (ML-RS), conventionally cooked (CL-RS), and autoclaved (AL-RS) lentil seeds. The size exclusion chromatography (SEC) results showed that UL-RS, RL-RS, and GL-RS (Group A samples) exhibited higher values of Mw and Rh¯ than FL-RS, ML-RS, AL-RS (Group C samples), and CL-RS (Group B sample). In parallel with the SEC result, other structural characteristics followed similar trends, where Group C samples exhibited the lowest values of double helix content and crystallinity by 13C NMR, and degree of order/double helix by FT-IR. Comparatively, Group A samples exhibited the opposite trends, and displayed large amorphous aggregates on their micrograph. The results are expected to provide information for better understanding the mechanism of resistant starch formation during different processing of lentil and to lay a theoretical foundation for the future study of their structure-function relationship.
Product Description
Supelco
Amyloglucosidase solution from Aspergillus niger, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
Diaminomaleonitrile, 98%
Sigma-Aldrich
Protease from Bacillus licheniformis, Type VIII, lyophilized powder, 7-15 units/mg solid