
TWEEN® 60

Synonym(s):
Polyethylene glycol sorbitan monostearate, Polyoxyethylene sorbitan monostearate
CAS Number:
Products
Product Category
Brand
Physical Form
Chemical Composition
Quality Segment
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing