
Retinyl palmitate

Synonym(s):
Vitamin A palmitate, all−trans−Retinol palmitate
Empirical Formula (Hill Notation):
C36H60O2
CAS Number:
Molecular Weight:
524.86
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3