
N-Benzyl-N-methylethanolamine

Synonym(s):
2-(N-Benzyl-N-methylamino)ethanol
Linear Formula:
C6H5CH2N(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
165.23
Products
Product Category
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Purity
Format
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing