
Linezolid Related Compound A

Synonym(s):
(5R)-5-(Azidomethyl)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxazolidinone, (R)-5-(Azidomethyl)-3-(3-fluoro-4-morpholinophenyl)oxazolidin-2-one
Empirical Formula (Hill Notation):
C14H16FN5O3
CAS Number:
Molecular Weight:
321.31
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2