
CPRG

Synonym(s):
Chlorophenol Red-β-D-galactopyranoside, CPRG, Chlorophenol Red-β-D-galactoside
Empirical Formula (Hill Notation):
C25H22Cl2O10S
CAS Number:
Molecular Weight:
585.41
Products
Formula Weight
Manufacturer
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing