
Clindamycin phosphate

Synonym(s):
Clindamycin 2-phosphate, Clindamycin 2-dihydrogen phosphate
Empirical Formula (Hill Notation):
C18H34ClN2O8PS
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3