
Chlorphenamine maleate

Synonym(s):
(±)-Chlorpheniramine maleate salt
Empirical Formula (Hill Notation):
C16H19ClN2 · C4H4O4
CAS Number:
Molecular Weight:
390.86
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing