
Calcium gluconate anhydrous

Synonym(s):
Calcium D-gluconate, 2,3,4,5,6-Pentahydroxycaproic Acid hemicalcium salt, Gluconic acid hemicalcium salt
Empirical Formula (Hill Notation):
C12H22CaO14
CAS Number:
Molecular Weight:
430.37
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing