
Bromophenol Blue solution

Synonym(s):
3′,3′′,5′,5′′-Tetrabromophenolsulfonephthalein, Bromophenol Blue sodium salt
Empirical Formula (Hill Notation):
C19H9Br4NaO5S
CAS Number:
Molecular Weight:
691.94
Products
Product Category
Brand
Formula Weight
pH Value
Physical Form
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing