
Anti-fd Bacteriophage antibody produced in rabbit

Synonym(s):
Anti-fd Bacteriophage
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Conjugate
Quality Segment
Package Size
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing