Skip to Content
Merck
Ammonium phosphomolybdate hydrate

Ammonium phosphomolybdate hydrate

Ammonium phosphomolybdate hydrate
Synonym(s):
Ammonium-12-molybdophosphate, Molybdophosphoric acid ammonium salt, Phosphomolybdic acid ammonium salt hydrate
Linear Formula:
(NH4)3PMo12O40 · xH2O
CAS Number:
Molecular Weight:
1876.35 (anhydrous basis)
EC Number:

Products

Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing