
Allopurinol Related Compound B

Synonym(s):
3-(Formylamino)-1H-pyrazole-4-carboxamide, 5-(Formylamino)-1H-pyrazole-4-carboxamide
Empirical Formula (Hill Notation):
C5H6N4O2
CAS Number:
Molecular Weight:
154.13
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2