Skip to Content
Merck
Adenosine 5′-triphosphate magnesium salt ≥95%, bacterial

Adenosine 5′-triphosphate magnesium salt

Adenosine 5′-triphosphate magnesium salt ≥95%, bacterial
Synonym(s):
ATP magnesium salt
Empirical Formula (Hill Notation):
C10H16N5O13P3 · xMg2+
CAS Number:
Molecular Weight:
507.18 (free acid basis)

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing