
5-Fluoro-L-tryptophan

Empirical Formula (Hill Notation):
C11H11FN2O2
CAS Number:
Molecular Weight:
222.22
Products
Brand
Sustainability
Color
Feature
Formula Weight
Manufacturer
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing