
3-(Perfluorobutyryl)-(+)-camphor

Synonym(s):
(+)-3-(Heptafluorobutyryl)-(+)-camphor, 3-(Perfluorobutyryl)-D-camphor
Empirical Formula (Hill Notation):
C14H15F7O2
Products
Product Category
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing