Skip to Content
Merck
All Photos(1)

Key Documents

PHL89543

Liquiritigenin

phyproof® Reference Substance

Synonym(s):

2,3-Dihydro-7-hydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 4′,7-Dihydroxyflavanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O4
CAS Number:
Molecular Weight:
256.25
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

InChI

1S/C15H12O4/c16-10-3-1-9(2-4-10)14-8-13(18)12-6-5-11(17)7-15(12)19-14/h1-7,14,16-17H,8H2/t14-/m0/s1

InChI key

FURUXTVZLHCCNA-AWEZNQCLSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Bioactive flavonoid from licorice exhibiting a neuroprotective effect.

Other Notes

This compound is commonly found in plants of the genus: glycyrrhiza

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Wang et al.
Biomedical chromatography : BMC, 34(4), e4788-e4788 (2020-01-04)
The objective of this paper was to develop a preparative method for the isolation and purification of liquiritigenin and glycyrrhetic acid from Glycyrrhiza uralensis Fisch using hydrolytic extraction combined with high-speed countercurrent chromatography (HSCCC). Liquiritigenin and glycyrrhetic acid were well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service