Merck
  • Competitive ELISA-like Label-free Detection of Lysozyme by Using a Fluorescent Monomer-doped Molecularly Imprinted Hydrogel.
Competitive ELISA-like Label-free Detection of Lysozyme by Using a Fluorescent Monomer-doped Molecularly Imprinted Hydrogel.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2017-11-14)
Takuya Kubo, Shunsuke Arimura, Toyohiro Naito, Tomoharu Sano, Koji Otsuka
ABSTRACT
We report on a molecularly imprinted hydrogel for the selective fluorescent detection of a targeting protein. To achieve the fluorescent detection of proteins, a fluorescent functional monomer was synthesized with fluorescein isothiocyanate and allylamine. The monomer (FITC-AA) provided a gradable alteration of the absorption spectrum, and a decrease of the fluorescent intensity due to an interaction with the model protein, lysozyme. Also, the imprinted hydrogel (PI-gel) was simply prepared with FITC-AA, the anionic functional monomers, poly(ethylene glycol) dimethacrylate, and lysozyme by radical photo polymerization. The PI-gel showed selective adsorption for lysozyme and the imprinting factor reached to 2.7. Additionally, the PI-gel indicated a gradual decrease of the fluorescent intensity corresponding to the lysozyme concertation similar to a competitive enzyme-linked immunosorbent assay detection.
Sigma-Aldrich
Sodium 4-vinylbenzenesulfonate, technical, ≥90% (T)
Sigma-Aldrich
2-Acrylamido-2-methyl-1-propanesulfonic acid, 99%