Merck
  • The additional effects of a probiotic mix on abdominal adiposity and antioxidant Status: A double-blind, randomized trial.
The additional effects of a probiotic mix on abdominal adiposity and antioxidant Status: A double-blind, randomized trial.
Obesity (Silver Spring, Md.) (2016-12-23)
Aline Corado Gomes, Rávila Graziany Machado de Sousa, Patrícia Borges Botelho, Tatyanne Letícia Nogueira Gomes, Patrícia Oliveira Prada, João Felipe Mota
ABSTRACT
To investigate whether a probiotic mix has additional effects when compared with an isolated dietary intervention on the body composition, lipid profile, endotoxemia, inflammation, and antioxidant profile. Women who had excess weight or obesity were recruited to a randomized, double-blind trial and received a probiotic mix (Lactobacillus acidophilus and casei; Lactococcus lactis; Bifidobacterium bifidum and lactis; 2 × 10 In comparison with the dietary intervention group, the dietary intervention + probiotic mix group showed a greater reduction in the waist circumference (-3.40% vs. -5.48%, P = 0.03), waist-height ratio (-3.27% vs. -5.00%, P = 0.02), conicity index (-2.43% vs. -4.09% P = 0.03), and plasma polyunsaturated fatty acids (5.65% vs. -18.63%, P = 0.04) and an increase in the activity of glutathione peroxidase (-16.67% vs. 15.62%, P < 0.01). Supplementation of a probiotic mix reduced abdominal adiposity and increased antioxidant enzyme activity in a more effective way than an isolated dietary intervention.
Product Description
Supelco
C4 - C24 Even Carbon Saturated FAMEs, 1000 μg/mL each component in hexane, analytical standard
Pricing and availability is not currently available.