Merck
  • Biological effects of double-walled carbon nanotubes on the innate immune system: An in vitro study on THP-1 human monocytes.
Biological effects of double-walled carbon nanotubes on the innate immune system: An in vitro study on THP-1 human monocytes.
Toxicology (2016-08-01)
Samir Dekali, Christine Bachelet, Séverine Maunoir-Regimbal, Emmanuel Flahaut, Jean-Claude Debouzy, David Crouzier
ABSTRACT
DWCNTs have numerous industrial and biomedical applications and several studies reported that they could act as immunomodulator systems. The immune system is the first line of defence of the human body when exposed to particulate matter. In order to investigate DWCNTs' role on innate immunity, we used THP-1 monocytic cells for the purpose of this study. We showed that DWCNTs were not cytotoxic until 6h, 24h, 48h and 72h of incubation with THP-1 monocytic cells (concentrations tested from 10 to 50μg/mL). From 6h to 72h of incubation of THP-1 cells with DWCNTs, we measured a significant increase of the baseline cell index using xCELLigence(®) technology showing cell adhesion. After 24h of exposure, DWCNTs agglomerates were localized in THP-1 monocyte cytoplasm and cell adhesion was observed simultaneously with a significant increase in the expression of CD11b and CD14 cell surface proteins. Pro-inflammatory cytokine secretion (IL-1β, IL-6, IL-8, TNF-α and IL-10) was also measured in supernatants after 6h or 24h of exposure to DWCNTs. This pro-inflammatory response was increased in THP-1 monocytic cells pre-treated with LPS. Altogether, our data indicate that DWCNTs induce an increased pro-inflammatory response of THP-1 monocytes and seem to modulate cell surface protein expression confirming that DWCNTs could act as stimulators of innate immunity.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Spurr Low Viscosity Embedding Kit, hydrophobic resin for embedding histological samples