Merck
  • Retrogradation enthalpy does not always reflect the retrogradation behavior of gelatinized starch.
Retrogradation enthalpy does not always reflect the retrogradation behavior of gelatinized starch.
Scientific reports (2016-02-11)
Shujun Wang, Caili Li, Xiu Zhang, Les Copeland, Shuo Wang
ABSTRACT
Starch retrogradation is a term used to define the process in which gelatinized starch undergoes a disorder-to-order transition. A thorough understanding of starch retrogradation behavior plays an important role in maintaining the quality of starchy foods during storage. By means of DSC, we have demonstrated for the first time that at low water contents, the enthalpy change of retrograded starch is higher than that of native starch. In terms of FTIR and Raman spectroscopic results, we showed that the molecular order of reheated retrograded starch samples is lower than that of DSC gelatinized starch. These findings have led us to conclude that enthalpy change of retrograded starch at low water contents involves the melting of recrystallized starch during storage and residual starch crystallites after DSC gelatinization, and that the endothermic transition of retrograded starch gels at low water contents does not fully represent the retrogradation behavior of starch. Very low or high water contents do not favor the occurrence of starch retrogradation.
Sigma-Aldrich
Amylose from potato, used as amylase substrate
Sigma-Aldrich
Amylopectin from potato starch