Merck
  • Lack of vitamin D receptor causes stress-induced premature senescence in vascular smooth muscle cells through enhanced local angiotensin-II signals.
Lack of vitamin D receptor causes stress-induced premature senescence in vascular smooth muscle cells through enhanced local angiotensin-II signals.
Atherosclerosis (2014-06-02)
Petya Valcheva, Anna Cardus, Sara Panizo, Eva Parisi, Milica Bozic, Jose M Lopez Novoa, Adriana Dusso, Elvira Fernández, Jose M Valdivielso
ABSTRACT
The inhibition of the renal renin-angiotensin system by the active form of vitamin D contributes to the cardiovascular health benefits of a normal vitamin D status. Local production of angiotensin-II in the vascular wall is a potent mediator of oxidative stress, prompting premature senescence. Herein, our objective was to examine the impact of defective vitamin D signalling on local angiotensin-II levels and arterial health. Primary cultures of aortic vascular smooth muscle cells (VSMC) from wild-type and vitamin D receptor-knockout (VDRKO) mice were used for the assessment of cell growth, angiotensin-II and superoxide anion production and expression levels of cathepsin D, angiotensin-II type 1 receptor and p57(Kip2). The in vitro findings were confirmed histologically in aortas from wild-type and VDRKO mice. VSMC from VDRKO mice produced more angiotensin-II in culture, and elicited higher levels of cathepsin D, an enzyme with renin-like activity, and angiotensin-II type 1 receptor, than wild-type mice. Accordingly, VDRKO VSMC showed higher intracellular superoxide anion production, which could be suppressed by cathepsin D, angiotensin-II type 1 receptor or NADPH oxidase antagonists. VDRKO cells presented higher levels of p57(Kip2), impaired proliferation and premature senescence, all of them blunted upon inhibition of angiotensin-II signalling. In vivo studies confirmed higher levels of cathepsin D, angiotensin-II type 1 receptor and p57(Kip2) in aortas from VDRKO mice. The beneficial effects of active vitamin D in vascular health could be a result of the attenuation of local production of angiotensin-II and downstream free radicals, thus preventing the premature senescence of VSMC.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-CTSD antibody produced in rabbit, affinity isolated antibody