Merck
  • AKI in early sepsis is a continuum from transient AKI without tubular damage over transient AKI with minor tubular damage to intrinsic AKI with severe tubular damage.
AKI in early sepsis is a continuum from transient AKI without tubular damage over transient AKI with minor tubular damage to intrinsic AKI with severe tubular damage.
International urology and nephrology (2014-09-06)
J Vanmassenhove, G Glorieux, E Hoste, A Dhondt, R Vanholder, W Van Biesen
ABSTRACT
The pathophysiology of septic acute kidney injury (AKI) is incompletely understood, and there is controversy on the role of renal hypoperfusion in early sepsis. We hypothesized that renal hypoperfusion plays a role in early sepsis and that there is a continuum between transient AKI without tubular damage, transient AKI with minor tubular damage, and intrinsic AKI. A total of 107 consecutive patients with sepsis were included. Fractional excretion of sodium (FENa), urinary, and serum neutrophil gelatinase-associated lipocalin were measured at admission (T0) and 4 h (T4) and 24 h later (T24). Patients were classified according to FENa quartiles (FENaQ). Transient and intrinsic AKI were respectively defined as AKI that did or did not recover to no AKI in the following 5 days. A total of 57 developed transient AKI, 22 developed intrinsic AKI, and 28 did not have AKI. Of the ten patients with transient AKI classified in the two lowest FENa quartiles (FENa < 0.36 %) and without signs of local tubular damage, seven still did not show signs of tubular damage 24 h later. Also, 50 % of patients with intrinsic AKI classified in the same FENaQ did not show signs of local tubular damage at admission but did so 24 h later. There is a continuum between transient AKI without tubular damage, transient AKI with minor tubular damage, and intrinsic AKI in sepsis. Renal hypoperfusion seems to be the instigator for the development of AKI in the majority of patients with early sepsis. Other mechanisms in some patients cannot be excluded.
Sigma-Aldrich
Mouse Lcn2 / Neutrophil Gelatinase-associated Lipocalin ELISA Kit
Pricing and availability is not currently available.
Sigma-Aldrich
Rat Lcn2 / Neutrophil Gelatinase-associated Lipocalin ELISA Kit
Pricing and availability is not currently available.
Sigma-Aldrich
Human Lipocalin-2 / NGAL ELISA Kit, for serum, plasma, cell culture supernatant and urine
Pricing and availability is not currently available.