Merck
  • In situ oligonucleotide synthesis on poly(dimethylsiloxane): a flexible substrate for microarray fabrication.
In situ oligonucleotide synthesis on poly(dimethylsiloxane): a flexible substrate for microarray fabrication.
Nucleic acids research (2005-05-05)
Matthew J Moorcroft, Wouter R A Meuleman, Steven G Latham, Thomas J Nicholls, Ryan D Egeland, Edwin M Southern
ABSTRACT
In this paper, we demonstrate in situ synthesis of oligonucleotide probes on poly(dimethylsiloxane) (PDMS) microchannels through use of conventional phosphoramidite chemistry. PDMS polymer was moulded into a series of microchannels using standard soft lithography (micro-moulding), with dimensions <100 microm. The surface of the PDMS was derivatized by exposure to ultraviolet/ozone followed by vapour phase deposition of glycidoxypropyltrimethoxysilane and reaction with poly(ethylene glycol) spacer, resulting in a reactive surface for oligonucleotide coupling. High, reproducible yields were achieved for both 6mer and 21mer probes as assessed by hybridization to fluorescent oligonucleotides. Oligonucleotide surface density was comparable with that obtained on glass substrates. These results suggest PDMS as a stable and flexible alternative to glass as a suitable substrate in the fabrication and synthesis of DNA microarrays.
Sigma-Aldrich
Iodine, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Pricing and availability is not currently available.