Merck
Telmisartan: just an antihypertensive agent? A literature review.
Expert opinion on pharmacotherapy (2011-11-15)
Maurizio Destro, Francesca Cagnoni, Giuseppina Patrizia Dognini, Vincenzo Galimberti, Carlo Taietti, Chiara Cavalleri, Emilio Galli
ABSTRACT
The modulation of the renin angiotensin aldosterone system (RAAS) is an important pathway in managing high blood pressure, and its overexpression plays a key role in target end-organ damage. Telmisartan is an angiotensin II receptor blocker (ARB) with unique pharmacologic properties, including the longest half-life among all ARBs; this leads to a significant and 24-h sustained reduction of blood pressure. Telmisartan has well-known antihypertensive properties, but there is also strong clinical evidence that it reduces left ventricular hypertrophy, arterial stiffness and the recurrence of atrial fibrillation, and confers renoprotection. This paper reviews telmisartan's pharmacological properties in terms of efficacy for hypertension control and, importantly, focuses on its new therapeutic indications and their clinical implications. ONTARGET (ongoing telmisartan alone and in combination with ramipril global endpoint trial) demonstrated, that telmisartan confers cardiovascular protective effects similar to those of ramipril, but with a better tolerability. Moreover, recent investigations focused on the capability of telmisartan to modulate the peroxisome proliferator-activated receptor-gamma (PPAR-γ), an established target in the treatment of insulin resistance, diabetes and metabolic syndrome, whose activation is also correlated to anti-inflammatory and, finally, anti-atherosclerotic properties. Telmisartan shows peculiar features that go beyond blood pressure control. It presents promising and unique protective properties against target end-organ damage, potentially able to open a scenario of new therapeutic approaches to cardiovascular disease.
Product Description
Telmisartan for peak identification, European Pharmacopoeia (EP) Reference Standard
Telmisartan for system suitability, European Pharmacopoeia (EP) Reference Standard
Telmisartan, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Telmisartan, ≥98% (HPLC), solid