Merck
  • Intramolecular silicon-assisted cross-coupling: total synthesis of (+)-brasilenyne.
Intramolecular silicon-assisted cross-coupling: total synthesis of (+)-brasilenyne.
Journal of the American Chemical Society (2002-12-19)
Scott E Denmark, Shyh-Ming Yang
ABSTRACT
The first, total synthesis of (+)-brasilenyne (1) has been achieved in 19 steps from l-(S)-malic acid. The key elements of this approach are a highly diastereoselective ring-opening of a 1,3-dioxolanone with bis(trimethylsilyl)acetylene) promoted by TiCl4 to set a propargylic stereocenter and the successful application of the sequential ring closing metathesis/silicon-assisted intramolecular cross-coupling reaction for construction of the oxonin core structure of 1.
Sigma-Aldrich
Bis(trimethylsilyl)acetylene, 99%
Pricing and availability is not currently available.