Merck
  • Farnesol is glucuronidated in human liver, kidney and intestine in vitro, and is a novel substrate for UGT2B7 and UGT1A1.
Farnesol is glucuronidated in human liver, kidney and intestine in vitro, and is a novel substrate for UGT2B7 and UGT1A1.
The Biochemical journal (2004-08-24)
Adam G Staines, Pavel Sindelar, Michael W H Coughtrie, Brian Burchell
ABSTRACT
Farnesol is an isoprenoid found in many aromatic plants and is also produced in humans, where it acts on numerous nuclear receptors and has received considerable attention due to its apparent anticancer properties. Although farnesol has been studied for over 30 years, its metabolism has not been well characterized. Recently, farnesol was shown to be metabolized by cytochromes P450 in rabbit; however, neither farnesol hydroxylation nor glucuronidation in humans have been reported to date. In the present paper, we show for the first time that farnesol is metabolized to farnesyl glucuronide, hydroxyfarnesol and hydroxyfarnesyl glucuronide by human tissue microsomes, and we identify the specific human UGTs (uridine diphosphoglucuronosyltransferases) involved. Farnesol metabolism was examined by a sensitive LC (liquid chromatography)-MS/MS method. Results indicate that farnesol is a good substrate for glucuronidation in human liver, kidney and intestine microsomes (values in nmol/min per mg). Initial analysis using expressed human UGTs indicated that UGTs 1A1 and 2B7 were primarily responsible for glucuronidation in vitro, with significantly lower activity for all the other UGTs tested (UGTs 1A3, 1A4, 1A6, 1A9 and 2B4). Kinetic analysis and inhibition experiments indicate that, in liver microsomes, UGT1A1 is primarily responsible for farnesol glucuronidation; however, in intestine microsomes, UGT2B7 is probably the major isoform involved, with a very-low-micromolar K(m). We also show the first direct evidence that farnesol can be metabolized to hydroxyfarnesol by human liver microsomes and that hydroxyfarnesol is metabolized further to hydroxyfarnesyl glucuronide. Thus glucuronidation may modulate the physiological and/or pharmacological properties of this potent signalling molecule.
Sigma-Aldrich
Farnesol, 95%
Sigma-Aldrich
Farnesol, mixture of isomers, ≥95%, stabilized, FG