Merck
  • Inhibition of acetaminophen and lorazepam glucuronidation in vitro by probenecid.
Inhibition of acetaminophen and lorazepam glucuronidation in vitro by probenecid.
Biopharmaceutics & drug disposition (1993-03-01)
L L von Moltke, M Manis, J S Harmatz, R Poorman, D J Greenblatt
ABSTRACT
The effect of probenecid on glucuronidation of acetaminophen and lorazepam in hepatic microsomes from various species was studied to see if in vitro results were consistent with previous in vivo observations. Mouse, rat, and human microsomes were incubated with acetaminophen and probenecid while monkey microsomes were incubated with lorazepam and probenecid. Glucuronidation rates in all species varied with substrate, protein, and detergent concentrations. Mice exhibited faster rates of glucuronidation than rats or humans. All species showed inhibition of glucuronidation of acetaminophen or lorazepam when probenecid was added. Analysis suggested competitive inhibition. Thus, in vitro studies support in vivo results and confirm that the inhibition takes place at the hepatic level.
Product Description
Supelco
Lorazepam glucuronide solution, 100 μg/mL in acetonitrile: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.