Merck
  • Quantitative determination of E- and Z-doxepin and E- and Z-desmethyldoxepin by high-performance liquid chromatography.
Quantitative determination of E- and Z-doxepin and E- and Z-desmethyldoxepin by high-performance liquid chromatography.
Therapeutic drug monitoring (1995-08-01)
M Adamczyk, J R Fishpaugh, C Harrington
ABSTRACT
A high-performance liquid chromatographic (HPLC) method was developed for the quantitative, simultaneous determination of the following four compounds in serum: E-doxepin, Z-doxepin, E-desmethyldoxepin, and Z-desmethyldoxepin. A 3-microns analytical silica column (6 x 100 mm) was employed with the mobile phase 0.025 M phosphate:acetonitrile:n-nonylamine (80:20:1). This HPLC method allows for the accurate measurement of all four isomeric compounds.
Product Description
Supelco
Nordoxepin hydrochloride, analytical standard, ≥98.0% (TLC), powder
Supelco
Desmethyldoxepin solution, cis/trans, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®