Merck
Membrane protein distribution in composite polymer-lipid thin films.
Chemical communications (Cambridge, England) (2012-07-28)
Johannes Thoma, Serena Belegrinou, Peggy Rossbach, Mariusz Grzelakowski, Katarzyna Kita-Tokarczyk, Wolfgang Meier
ABSTRACT
We present a model system to demonstrate that the positioning of biomolecules (membrane proteins) in a nonnative, complex thin film environment can be regulated by the phase behavior of film components. Partial separation between an amphiphilic polymer and a lipid drives the protein to a fluid phase, mechanically more similar to a cellular bilayer.
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, ≥99% (TLC)
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, semisynthetic, ≥99%