Merck
  • Bonding to NaOCl-treated dentin: effect of pretreatment with sodium toluene sulfinic acid.
Bonding to NaOCl-treated dentin: effect of pretreatment with sodium toluene sulfinic acid.
The journal of adhesive dentistry (2011-09-22)
Xianghong Li, Weidong Ma, Xiaodong Chen, Jiaming Zhao, Takashi Nakata, Kumiko Tanaka, Kei Takahashi, Yoshihiro Nishitani, Masahiro Yoshiyama
ABSTRACT
The use of sodium hypochlorite (NaOCl) can decrease the bond strength of 4-META/MMA-TBB resin cement to root dentin. The aim of this in vitro study was to evaluate the effect of sodium toluene sulfinic acid (SA) as a pretreatment to increase bond strength to NaOCl-treated dentin. The root canal dentin of bovine teeth was treated with 6 methods: group 10-3 (10% citric acid/3% ferric chloride; control); group SA; group NC (sodium hypochlorite/10-3 solution); group NS-10-3 (NaOCl + sulfinic acid + 10-3 solution); group HSA-10-3 (NaOCl +H2O2 +NaOCl + sulfinic acid + 10-3 solution); group HO-10-3 (NaOCl + H2O2 + NaOCl +10-3 solution). The roots were then filled with Super-Bond C&B (SB) or Super-Bond sealer (SBS). Samples were stored in distilled water for 24 h at 37°C and then cross sectioned into five slabs (0.8 mm thick) that were subjected to "trimming" microtensile bond strength testing. All treated dentin surfaces and resin/dentin bond interfaces were analyzed under SEM. Compared with the control, NaOCl treatment significantly decreased the resin/dentin bond strengths. However, SA treatment following NaOCl irrigation resulted in no significant differences of bond strength values. Sulfinic acid was effective in restoring 4-META/MMA-TBB resin cement bond strength after NaOCl irrigation.
Sigma-Aldrich
Sodium p-toluenesulfinate, 95%
Sigma-Aldrich
Sodium p-toluenesulfinate, purum, anhydrous, ≥96.0% (NT)