Merck
  • Potentiometric polymeric membrane electrodes for mercury detection using calixarene ionophores.
Potentiometric polymeric membrane electrodes for mercury detection using calixarene ionophores.
Water science and technology : a journal of the International Association on Water Pollution Research (2010-02-13)
Sonika Tyagi, Himanshu Agarwal, Saiqa Ikram
ABSTRACT
It is here established that potentiometric polymeric membrane electrodes based on electrically neutral ionophores are a useful analytical tool for the detection of heavy metal ions from environmental and industrial waste water. PVC based membrane containing p-tert-butyl-calix[4]arenethioether derivative as active material along with sodiumtetraphenylborate (NaTPB) as solvent mediator and dibutylphthalate as a plasticizer in the ratio 45:9:460:310 (w/w%) (I:NaTPB:DBP:PVC) exhibits good properties with a Nernstian response of 29.50+/-1.0 mV per decade of activity and a working concentration range of 7.2 x 10(-8)-1.0 x 10(-1) M. The electrode gave more stable potential readings when used around pH 2.5-6.8 and exhibits fast response time of 14 s. The sensors were found to work satisfactorily in partially non-aqueous media up to 40% (v/v) content of acetone, methanol or ethanol and could be used over a period of 7-9 months. Excellent selectivity for Hg(2+) ions is indicated by match potential method and fixed interference method. The sensors could be used successfully in the estimation of mercury in different sample.
Sigma-Aldrich
Potassium tetraphenylborate, 97%
Sigma-Aldrich
Ammonium tetraphenylborate, 99%
Sigma-Aldrich
Sodium tetraphenylborate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium tetraphenylborate, puriss. p.a., ACS reagent, ≥99.5% (NT)