Merck
Intermolecular hydroaminations via strained (E)-cycloalkenes.
The Journal of organic chemistry (2007-12-29)
Joseph Moran, Pamela H Cebrowski, André M Beauchemin
ABSTRACT
A photoinduced procedure for the intermolecular hydroamination of alkenes using azoles is described. This reaction occurs in modest to good yield for 6- and 7-membered cyclic alkenes. Upon irradiation at 254 nm in the presence of methyl benzoate and a small amount of triflic acid as an additive (20 mol %), imidazoles, pyrazoles, triazoles, and tetrazole can react with the alkene to afford complex Markovnikov adducts. The proposed mechanism involves photoisomerization to generate highly strained (E)-cycloalkene intermediates and (E)-cycloalkene protonation followed by reaction with the azole nucleophile. Alkene isomerization was found to be a competing side reaction under these conditions.
Product Description
Supelco
Methyl benzoate, analytical standard
Sigma-Aldrich
Methyl benzoate, 99%
Sigma-Aldrich
Methyl benzoate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Methyl benzoate, ≥98%, FCC, FG