Merck
  • Evaluation of pirimiphos-methyl (50% EC) against the immatures of Anopheles stephensi/An. culicifacies (malaria vectors) and Culex quinquefasciatus (vector of bancroftian filariasis).
Evaluation of pirimiphos-methyl (50% EC) against the immatures of Anopheles stephensi/An. culicifacies (malaria vectors) and Culex quinquefasciatus (vector of bancroftian filariasis).
Journal of vector borne diseases (2004-08-31)
M A Ansari, P K Mittal, R K Razdan, R C Dhiman, A Kumar
ABSTRACT
In India, temephos and fenthion are used as larvicides in fresh and polluted waters. Since use of same insecticide may precipitate resistant, as an alternative bioefficacy of pirimiphos-methyl--an organophosphorus insecticide was evaluated against immatures of Anopheles and Culex species in different breeding habitats in District Ghaziabad (U.P.) and Goa. Laboratory bioassays were carried out using standard WHO procedure. LC50 and LC90 were calculated using Probit analysis. Abbott's formula was used to calculate per cent corrected mortality in laboratory exposures while, Mulla's formula was applied for calculating per cent reduction in mosquito immatures under field conditions. Pirimiphos-methyl was found to be most effective against larvae of An. stephensi followed by An. culicifacies and Cx. quinquefasciatus. The LC50 and LC90 values against three species were 0.023, 0.032 and 0.04 ppm; and 0.045, 0.057 and 0.114 ppm respectively. In field, pirimiphos-methyl @ 200 g a.i./ha against Anopheles species and @ 300 g a.i./ha against Culex species can produce > 80% reduction in density of immatures up to one week in different habitats. Pirimiphos-methyl < 0.25 ppm is not toxic to fish. Pirimiphos-methyl is more effective against anophelines in clean water than culicines in polluted water. This larvicide should not be used at a concentration above 0.25 ppm in habitats harbouring the larvivorous fish.
Product Description
Supelco
Pirimiphos-methyl, PESTANAL®, analytical standard