Merck
  • Characterization of a strain of Sphingobacterium sp. and its degradation to herbicide mefenacet.
Characterization of a strain of Sphingobacterium sp. and its degradation to herbicide mefenacet.
Journal of environmental sciences (China) (2004-05-13)
Yang-fang Ye, Hang Min, Yu-feng Du
ABSTRACT
A bacterium (designated strain Y1) degrading acetanilide herbicide mefenacet was isolated from aerobic sludge. Based on the analyses of partial 16S rRNA gene, cellular fatty acid and BIOLOG-GN, and general physiological and biochemical characteristics, strain Y1 was identified as Sphingobacterium multivolum. Strain Y1 was able to degrade mefenacet used as sources of carbon and energy. Degradation of mefenacet was accompanied by producing the metabolites N-methylaniline and an unidentified compound with molecular weight 205, indicating a metabolic pathway of mefenacet initiated by hydrolysis of amido bond.
Sigma-Aldrich
N-Methylaniline, ≥99%
Sigma-Aldrich
N-Methylaniline, 98%
Supelco
N-Methylaniline, analytical standard
Sigma-Aldrich
N-Methylaniline, purum, ≥98.0% (GC)