Merck
  • Serine and threonine beta-lactones: a new class of hepatitis A virus 3C cysteine proteinase inhibitors.
Serine and threonine beta-lactones: a new class of hepatitis A virus 3C cysteine proteinase inhibitors.
The Journal of organic chemistry (2002-03-02)
Manjinder S Lall, Yeeman K Ramtohul, Michael N G James, John C Vederas
ABSTRACT
Hepatitis A virus (HAV) 3C enzyme is a cysteine proteinase essential for viral replication and infectivity and represents a target for the development of antiviral drugs. A number of serine and threonine beta-lactones were synthesized and tested against HAV 3C proteinase. The D-N-Cbz-serine beta-lactone 5a displays competitive reversible inhibition with a K(i) value of 1.50 x 10(-6) M. Its enantiomer, L-N-Cbz-serine beta-lactone 5b is an irreversible inactivator with k(inact) = 0.70 min(-1), K(Iota) = 1.84 x 10(-4) M and k(inact)/K(Iota) = 3800 M(-1) min(-1). Mass spectrometry and HMQC NMR studies using (13)C-labeled 5b show that inactivation of the enzyme occurs by nucleophilic attack of the cysteine thiol (Cys-172) at the beta-position of the oxetanone ring. Although the N-Cbz-serine beta-lactones 5a and 5b display potent inhibition, other related analogues with an N-Cbz side chain, such as the five-membered ring homoserine gamma-lactones 14a and 14b, the four-membered ring beta-lactam 33, 2-methylene oxetane 34, cyclobutanone 36, and 3-azetidinone 39, fail to give significant inhibition of HAV 3C proteinase, thus demonstrating the importance of the beta-lactone ring for binding.
Sigma-Aldrich
Hymeglusin, ≥98% (HPLC), from Fusarium sp.