Merck
  • Gene expression profiling in human esophageal cancers using cDNA microarray.
Gene expression profiling in human esophageal cancers using cDNA microarray.
Biochemical and biophysical research communications (2001-08-25)
T Kan, Y Shimada, F Sato, M Maeda, A Kawabe, J Kaganoi, A Itami, S Yamasaki, M Imamura
ABSTRACT
Human esophageal cancer cell lines and human esophageal cancer tissues were profiled on cDNA microarrays. In esophageal cancer cell lines, KYAE and OE-33 (adenocarcinomas) were distinguished from KYSE series (squamous cell carcinomas). Although SK-GT-4 and TE7 were derived from adenocarcinomas, they had a comparatively similar expression profile to the KYSE series. A set of genes whose expression commonly either increased or decreased in cancer cell lines was identified. Genes that were characteristically expressed in KYAE and OE-33 were also identified. The gene expression profiles of cancer tissues (CTs) were remarkably different from those of the cancer cell lines (CCLs). Notable differences between CCLs and CTs were observed in matrix metalloproteinases, plasminogen activator, collagens, paxillin, and thrombospondin 2, etc., whose expression was not increased in CCLs but increased in CTs. Twenty-three genes were extracted to categorize patients according to their prognoses, and clustering analyses, using these genes, were performed successfully.
Sigma-Aldrich
KYAE-1 cell line, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.