Merck
  • Exosome inhibition improves response to first-line therapy in small cell lung cancer.
Exosome inhibition improves response to first-line therapy in small cell lung cancer.
Journal of cellular and molecular medicine (2024-02-14)
Nesrin Irep, Kubilay Inci, Pervin Elvan Tokgun, Onur Tokgun
ABSTRACT
Exosomes are recognized as important mediators of cell-to-cell communication, facilitating carcinogenesis. Although there have been significant advancements in exosome research in recent decades, no drugs that target the inhibition of sEV secretion have been approved for human use. For this study, we employed GW4869 and Nexinhib20 as inhibitors of exosome synthesis and trafficking combined. First, we found that Nexinhib20 and GW4869 effectively inhibited RAB27A and neutral sphingomyelinase 2 (nSMase2) nsMase2. Interestingly, the inhibition of nsMase2 and RAB27A decreased expression of CD9, CD63 and Tsg101, both at RNA and protein levels. We used a combination treatment strategy of cisplatin/etoposide plus GW4869 or Nexinhib20 on small cell lung cancer (SCLC) cell lines. The combination treatment of GW4869 or Nexinhib20 effectively enhanced the inhibitory effects of first-line chemotherapy on the SCLC cells. Furthermore, we demonstrated that reducing exosome release through GW4869 and Nexinhib20 treatment effectively reduced cellular proliferation and significantly induced apoptosis in SCLC cells. Also, we showed that combining exosome inhibition with chemotherapy has a significant synergistic effect on cellular proliferation. We also found increased p53 and p21 expressions with western blot and significantly changing Bax, BCL2, caspase-3 and caspase-9 expressions. Inhibiting the exosome pathway offers opportunities for developing novel, effective treatment strategies for SCLC.
Sigma-Aldrich
Nexinhib20, ≥98% (HPLC)
Pricing and availability is not currently available. Contact Technical Service